O412 98%

Supplier: Ambeed
Warning

Synonyms: 2-[(4-chlorophenyl)amino]- 4-thiazolecarboxylic acid, ethyl, Ethyl 2-((4-chlorophenyl)amino)thiazole-4-carboxylate, O4I2

A221899-10MG A221899-1MG A221899-25MG A221899-50MG A221899-5MG A221899-100MG A221899-250MG
76800-888EA 38.2 USD
76800-888 76800-890 76800-892 76800-894 76800-896 77667-844 77667-846
O412 98%
O412

Precaución: for Research Use Only

Formula: C₁₂H₁₁ClN₂O₂S
MW: 282.70 g/mol
MDL Number: MFCD08166530
CAS Number: 165682-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR