5-(Chloroacetyl)oxindole 98%

Supplier: Ambeed
Danger

A220492-10G A220492-1G A220492-25G A220492-250MG
76800-876EA 41 USD
76800-876 76800-878 77879-349 77879-348
5-(Chloroacetyl)oxindole 98%
5-(Chloroacetyl)oxindole

Precaución: for Research Use Only

Formula: C₁₀H₈ClNO₂
MDL Number: MFCD04115734
CAS Number: 65435-04-3
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR