Glycolaldehyde dimethyl acetal 96%

Supplier: Ambeed
Warning

Synonyms: 2,2-Dimethoxyethanol

A220108-1G A220108-25G A220108-5G A220108-100G
76800-870EA 19 USD
76800-870 76800-872 76800-874 77879-344
Glycolaldehyde dimethyl acetal 96%
Glycolaldehyde dimethyl acetal

Precaución: for Research Use Only

Formula: HOCH₂CH(OCH₃)₂
MDL Number: MFCD00051799
CAS Number: 30934-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR