Diethylene glycol dimethacrylate 98% stabilized
Supplier: Ambeed
|
Warning
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ) 2-3%
A219691-1000G
A219691-100G
A219691-25G
A219691-500G
76800-860EA
102
USD
76800-860
76800-862
76800-864
76800-866
Diethylene glycol dimethacrylate 98% stabilized
Diethylene glycol dimethacrylate
Precaución: for Research Use Only
Formula:
C₁₂H₁₈O₅ |
MDL Number:
MFCD00014931 CAS Number: 2358-84-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...