5-Chloro-2-phenoxyaniline 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-chlorodiphenyl ether, 2-Amino-4-chlorophenyl phenyl ether, 5-Chloro-2-phenoxyaniline, 2-Amino-4-chloro-diphenyl ether

A219024-100G A219024-25G A219024-5G
76800-840EA 117.76 USD
76800-840 76800-842 76800-844
5-Chloro-2-phenoxyaniline 97%
5-Chloro-2-phenoxyaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClNO
MW: 219.6700 g/mol
MDL Number: MFCD00025217
CAS Number: 93-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR