Methyl (2,5-dimethyl-1,3-thiazol-4-yl)acetate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(2,5-dimethylthiazol-4-yl)acetate

A218730-1G A218730-250MG
76800-830EA 65.78 USD
76800-830 76800-832
Methyl (2,5-dimethyl-1,3-thiazol-4-yl)acetate 97%
Methyl (2,5-dimethyl-1,3-thiazol-4-yl)acetate

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂S
MW: 185.243 g/mol
MDL Number: MFCD02677686
CAS Number: 306937-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR