(S)-Methyl-6-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-aminohexanoate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-Methyl 6-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-aminohexanoate hydrochloride, H-Lys(Fmoc)-OMe·HCl
A218519-1G
A218519-250MG
A218519-25G
A218519-5G
A218519-10G
76800-820EA
6
USD
76800-820
76800-822
76800-824
76800-826
77879-335
(S)-Methyl-6-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-aminohexanoate hydrochloride 97%
(S)-Methyl-6-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-aminohexanoate hydrochloride
Precaución: for Research Use Only
Formula:
C₂₂H₂₇ClN₂O₄ |
MDL Number:
MFCD00153447 CAS Number: 201009-98-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...