2-Bromo-6-methoxynaphthalene 98%

Supplier: Ambeed
Warning

A218516-100G A218516-10G A218516-25G A218516-500G
76800-814EA 38.2 USD
76800-814 76800-816 76800-818 77600-762
2-Bromo-6-methoxynaphthalene 98%
2-Bromo-6-methoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₁H₉BrO
MW: 237.1 g/mol
MDL Number: MFCD00004062
CAS Number: 5111-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR