(S)-N-Boc-4-thiazoylalanine 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-3-(thiazol-4-yl)propanoic acid, Boc-β-(4-Thiazolyl)-Ala-OH

A217955-250MG A217955-1G A217955-5G
76800-810EA 31 USD
76800-810 77568-114 77879-334
(S)-N-Boc-4-thiazoylalanine 98%
(S)-N-Boc-4-thiazoylalanine

Precaución: For research use only.

Formula: C₁₁H₁₆N₂O₄S
MDL Number: MFCD00079682
CAS Number: 119434-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR