Eprosartan Mesylate 99%

Supplier: Ambeed
Warning

A217175-100MG A217175-10MG A217175-50MG A217175-250MG A217175-1G
76800-792EA 10 USD
76800-792 76800-794 76800-796 77626-102 77879-332
Eprosartan Mesylate 99%
Eprosartan Mesylate

Precaución: for Research Use Only

Formula: C₂₃H₂₄N₂O₄S·CH₄O₃S
MDL Number: MFCD08141807
CAS Number: 144143-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR