Fmoc-3-carboxypiperidine 97%

Supplier: Ambeed
Warning

Synonyms: 1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidine-3-carboxylic acid

A216420-1G A216420-25G A216420-5G
76800-770EA 18 USD
76800-770 76800-772 76800-774
Fmoc-3-carboxypiperidine 97%
Fmoc-3-carboxypiperidine

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₄
MDL Number: MFCD00673789
CAS Number: 158922-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR