Ethyl-7-nitroindole-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 7-nitro-1H-indole-2-carboxylate

A216340-100G A216340-10G A216340-25G A216340-5G
76800-762EA 1472.53 USD
76800-762 76800-764 76800-766 76800-768
Ethyl-7-nitroindole-2-carboxylate 97%
Ethyl-7-nitroindole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O₄
MW: 234.211 g/mol
MDL Number: MFCD00216475
CAS Number: 6960-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR