all cis-5,8,11,14,17-Eicosapentaenoic acid 95%

Supplier: Ambeed
Danger

Synonyms: 5(Z),8(Z),11(Z),14(Z),17(Z)-Eicosapentaenoic Acid, Timnodonic acid, cis-5,8,11,14,17-Eicosapentaenoic acid, EPA

A216225-25G A216225-5G A216225-1G
76800-760EA 143 USD
76800-760 77550-398 77568-088
all cis-5,8,11,14,17-Eicosapentaenoic acid 95%
all cis-5,8,11,14,17-Eicosapentaenoic acid

Precaución: For research use only.

Formula: C₂₀H₃₀O₂
MW: 302.46 g/mol
MDL Number: MFCD00065716
CAS Number: 10417-94-4
UN: 3265
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR