Pamoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Methylenebis(3-hydroxy-2-naphthoic acid), Embonic acid

A215829-100G A215829-10G A215829-5G A215829-25G
76800-752EA 24.41 USD
76800-752 76800-754 76800-756 77626-084
Pamoic acid 97%
Pamoic acid

Precaución: for Research Use Only

Formula: C₂₃H₁₆O₆
MW: 388.38 g/mol
MDL Number: MFCD00004079
CAS Number: 130-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR