4-(Bis(2-hydroxyethyl)amino)benzaldehyde 95%

Supplier: Ambeed
Warning

A215718-100MG A215718-250MG
76800-750EA 35.01 USD
76800-750 77667-806
4-(Bis(2-hydroxyethyl)amino)benzaldehyde 95%
4-(Bis(2-hydroxyethyl)amino)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₃
MW: 209.24 g/mol
MDL Number: MFCD04114397
CAS Number: 27913-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR