Ethyl-p-anisate 95%

Supplier: Ambeed
Warning

Synonyms: p-Anisic acid ethyl ester, Ethyl 4-methoxybenzoate, 4-Methoxybenzoic acid ethyl ester, 4-Anisic acid ethyl ester

A214928-10G A214928-25G
76800-734EA 7.43 USD
76800-734 76800-736
Ethyl-p-anisate 95%
Ethyl-p-anisate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
MDL Number: MFCD00015151
CAS Number: 94-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR