Dimethyl 2-bromopentanedioate 95%

Supplier: Ambeed
Warning

A214501-100MG A214501-250MG A214501-1G
76800-730EA 16 USD
76800-730 76800-732 77550-378
Dimethyl 2-bromopentanedioate 95%
Dimethyl 2-bromopentanedioate

Precaución: for Research Use Only

Formula: C₇H₁₁BrO₄
MDL Number: MFCD00043630
CAS Number: 760-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR