Methyl-2,4-dimethoxybenzoate 98%

Supplier: Ambeed
Warning

A214458-25G A214458-5G
76800-726EA 85 USD
76800-726 76800-728
Methyl-2,4-dimethoxybenzoate 98%
Methyl-2,4-dimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MDL Number: MFCD00070649
CAS Number: 2150-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR