Diethyl (2,2-diethoxyethyl)phosphonate 98%

Supplier: Ambeed
Warning

Synonyms: Diethyl 2,2-diethoxy-ethanephosphonate

A214388-25G A214388-5G A214388-1G
76800-724EA 170.81 USD
76800-724 77568-066 77626-080
Diethyl (2,2-diethoxyethyl)phosphonate 98%
Diethyl (2,2-diethoxyethyl)phosphonate

Precaución: For research use only.

Formula: C₁₀H₂₃O₅P
MW: 254.26 g/mol
MDL Number: MFCD00009236
CAS Number: 7598-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR