4-Cyano-3-fluorophenyl 4-Propylbenzoate 97%

Supplier: Ambeed
Warning

A212854-1G A212854-5G A212854-25G
76800-686EA 9 USD
76800-686 76800-688 77879-315
4-Cyano-3-fluorophenyl 4-Propylbenzoate 97%
4-Cyano-3-fluorophenyl 4-Propylbenzoate

Precaución: for Research Use Only

Formula: C₁₇H₁₄FNO₂
MDL Number: MFCD06797522
CAS Number: 86776-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR