Chromone-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A212074-1G A212074-250MG A212074-5G A212074-25G
76800-662EA 5.31 USD
76800-662 76800-664 76800-666 76800-668 77879-309
Chromone-3-carboxylic acid 95%
Chromone-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆O₄
MDL Number: MFCD00017338
CAS Number: 39079-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR