Fmoc-D-Thr(Bzl)-OH 95%

Supplier: Ambeed
Warning

Synonyms: (2R,3S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(benzyloxy)butanoic acid

A210728-1G A210728-250MG A210728-5G
76800-630EA 23.34 USD
76800-630 76800-632 76800-634
Fmoc-D-Thr(Bzl)-OH 95%
Fmoc-D-Thr(Bzl)-OH

Precaución: for Research Use Only

Formula: C₂₆H₂₅NO₅
MW: 431.488 g/mol
MDL Number: MFCD00237033
CAS Number: 131545-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR