O-(Furan-2-ylmethyl) ethanethioate 97%

Supplier: Ambeed
Warning

A209849-100G A209849-25G A209849-500G
76800-610EA 37.14 USD
76800-610 76800-612 76800-614
O-(Furan-2-ylmethyl) ethanethioate 97%
O-(Furan-2-ylmethyl) ethanethioate

Precaución: for Research Use Only

Formula: C₇H₈O₂S
MW: 156.20 g/mol
MDL Number: MFCD00010083
CAS Number: 125942-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR