Diphenyl sulfone 98%

Supplier: Ambeed
Warning

Synonyms: Phenyl sulphone, sulphonyldibenzene

A209607-1000G A209607-100G A209607-500G
76800-598EA 35.01 USD
76800-598 76800-600 76800-602
Diphenyl sulfone 98%
Diphenyl sulphone

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂S
MW: 218.28 g/mol
MDL Number: MFCD00007548
CAS Number: 127-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR