indane-5-sulfonyl chloride 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2,3-Dihydro-1H-indene-5-sulphonyl chloride
A209168-100MG
A209168-1G
A209168-250MG
A209168-5G
76800-574EA
6
USD
76800-574
76800-576
76800-578
76800-580
indane-5-sulfonyl chloride 98%
indane-5-sulfonyl chloride
Precaución: for Research Use Only
Formula:
C₉H₉ClO₂S MW: 216.679 g/mol |
MDL Number:
MFCD05237217 CAS Number: 52205-85-3 UN: 3261 ADR: 8 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...