Lapatinib ditosylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tyverb, N-(3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)-6-(5-(((2-(methylsulphonyl)ethyl)amino)methyl)furan-2-yl)quinazolin-4-amine bis(4-methylbenzenesulphonate), Tykerb, N-(3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)-6-(5-(((2-(methylsulfonyl)ethyl)amino)methyl)furan-2-yl)quinazolin-4-amine bis(4-methylbenzenesulphonate), GW572016 ditosylate, GW2016 ditosylate
A208985-50MG
A208985-25G
A208985-250MG
A208985-5G
A208985-1G
76800-556EA
6
USD
76800-556
77568-006
77667-760
77667-762
77879-301
Lapatinib ditosylate 98%
Lapatinib ditosylate
Precaución: For research use only.
Formula:
C₂₉H₂₆ClFN₄O₄S·₂C₇H₈O₃S MW: 925.46 g/mol |
MDL Number:
MFCD09264195 CAS Number: 388082-77-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...