Glyoxal dimethyl acetal 60% (w/w) in water

Supplier: Ambeed
Warning

Synonyms: Dimethoxyacetaldehyde, 2,2-Dimethoxyacetaldehyde, Dimethoxyethanal

A208718-5G A208718-10G A208718-25G A208718-500G A208718-100G
76800-554EA 6 USD
76800-554 77550-344 77550-346 77568-000 77600-686
Glyoxal dimethyl acetal 60% (w/w) in water
Glyoxal dimethyl acetal

Precaución: For research use only.

Formula: OHCCH(OCH₃)₂
MDL Number: MFCD00134410
CAS Number: 51673-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR