2,3-Dihydro-1H-indol-4-amine 95%

Supplier: Ambeed
Warning

Synonyms: Indolin-4-amine

A207314-100MG A207314-1G A207314-250MG
76800-538EA 76.39 USD
76800-538 76800-540 76800-542
2,3-Dihydro-1H-indol-4-amine 95%
2,3-Dihydro-1H-indol-4-amine

Precaución: for Research Use Only

Formula: C₈H₁₀N₂
MW: 134.178 g/mol
MDL Number: MFCD08234591
CAS Number: 52537-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR