DL-Homocysteic acid 96%

Supplier: Ambeed
Warning

A207039-10G A207039-1G A207039-25G A207039-5G
76800-530EA 100 USD
76800-530 76800-532 76800-534 76800-536
DL-Homocysteic acid 96%
DL-Homocysteic acid

Precaución: for Research Use Only

Formula: C₄H₉NO₅S
MDL Number: MFCD00007536
CAS Number: 504-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR