1-Amino-2-methylnaphthalene 95%

Supplier: Ambeed
Warning

Synonyms: 2-Methylnaphthalen-1-amine, 2-Methyl-1-naphthylamine

A206687-100G A206687-1G A206687-250MG A206687-25G A206687-5G
76800-520EA 754.3 USD
76800-520 76800-522 76800-524 76800-526 76800-528
1-Amino-2-methylnaphthalene 95%
1-Amino-2-methylnaphthalene

Precaución: for Research Use Only

Formula: C₁₁H₁₁N
MW: 157.22 g/mol
MDL Number: MFCD00059135
CAS Number: 2246-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR