4-Acetoxyindole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Indolyl acetate

A206293-100G A206293-10G A206293-1G A206293-25G A206293-5G A206293-500G
76800-508EA 665.19 USD
76800-508 76800-510 76800-512 76800-514 76800-516 77550-320
4-Acetoxyindole 98%
4-Acetoxyindole

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
MDL Number: MFCD00010678
CAS Number: 5585-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR