Isoquinoline-3-carboxylic acid monohydrate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Azanaphthalene-3-carboxylic acid monohydrate, 3-Carboxyisoquinoline monohydrate, Isoquinoline-3-carboxylic acid hydrate
A205967-1G
A205967-5G
76800-504EA
18.04
USD
76800-504
76800-506
Isoquinoline-3-carboxylic acid monohydrate 95%
Isoquinoline-3-carboxylic acid monohydrate
Precaución: for Research Use Only
Formula:
C₁₀H₉NO₃ MW: 191.183 g/mol |
MDL Number:
MFCD00167216 CAS Number: 207399-25-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...