2-(Benzyl(phenyl)amino)ethyl 5-(5,5-dimethyl-2-oxido-1,3,2-dioxaphosphinan-2-yl)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3-carboxylate ethanol hydrochloride 98%

Supplier: Ambeed
Warning

A205020-10MG A205020-1MG A205020-5MG
76800-490EA 91 USD
76800-490 76800-492 76800-494
2-(Benzyl(phenyl)amino)ethyl 5-(5,5-dimethyl-2-oxido-1,3,2-dioxaphosphinan-2-yl)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3-carboxylate ethanol hydrochloride 98%
2-(Benzyl(phenyl)amino)ethyl 5-(5,5-dimethyl-2-oxido-1,3,2-dioxaphosphinan-2-yl)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3-carboxylate ethanol hydrochloride

Precaución: for Research Use Only

Formula: C₃₆H₄₅ClN₃O₈P
MDL Number: MFCD00875717
CAS Number: 111011-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR