Fmoc-Lys(Me3)-OH chloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-Lys(Me)₃-OH chloride, Fmoc-Lys(Me)3-OH Chloride, Fmoc-Lys(Me)3-OH Chloride
A204695-100MG
A204695-1G
A204695-250MG
A204695-5G
76800-482EA
38
USD
76800-482
76800-484
76800-486
76800-488
Fmoc-Lys(Me3)-OH chloride 97%
Fmoc-Lys(Me3)-OH chloride
Precaución: for Research Use Only
Formula:
C₂₄H₃₁ClN₂O₄ MW: 446.972 g/mol |
MDL Number:
MFCD00672333 CAS Number: 201004-29-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...