D-(+)-Maltose 95%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, Maltose

A204105-100G A204105-10G A204105-25G A204105-1000G A204105-500G
76800-460EA 43 USD
76800-460 76800-462 76800-464 77600-666 77600-668
D-(+)-Maltose 95%
D-(+)-Maltose

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₁₁
MDL Number: MFCD00135877
CAS Number: 69-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR