Iron(III) p-toluenesulfonate 98%
Supplier: Ambeed
|
Warning
|
A203340-100G
A203340-25G
A203340-5G
A203340-1000G
A203340-500G
76800-436EA
18
USD
76800-436
76800-438
76800-440
77667-694
77667-696
Iron(III) p-toluenesulfonate 98%
Iron(III) p-toluenesulfonate
Precaución: for Research Use Only
Formula:
(C₇H₇O₃S)₃·Fe |
MDL Number:
MFCD01766290 CAS Number: 77214-82-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...