2-Amino-2-(hydroxymethyl)propane-1,3-diol hemi(2,2'-((2-chloro-5-cyano-1,3-phenylene)bis(azanediyl))bis(2-oxoacetate)) 95%

Supplier: Ambeed
Danger

Synonyms: Lodoxamide tromethamine

A202923-10MG A202923-1MG A202923-25MG A202923-50MG A202923-5MG
76800-406EA 15.92 USD
76800-406 76800-408 76800-410 76800-412 76800-414
2-Amino-2-(hydroxymethyl)propane-1,3-diol hemi(2,2'-((2-chloro-5-cyano-1,3-phenylene)bis(azanediyl))bis(2-oxoacetate)) 95%
2-Amino-2-(hydroxymethyl)propane-1,3-diol hemi(2,2'-((2-chloro-5-cyano-1,3-phenylene)bis(azanediyl))bis(2-oxoacetate))

Precaución: for Research Use Only

Formula: C₁₉H₂₈ClN₅O₁₂
MW: 553.91 g/mol
MDL Number: MFCD04112765
CAS Number: 63610-09-3
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR