2-Chloro-6-methylquinoline-3-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-6-methylquinoline-3-carboxaldehyde, 2-Chloro-3-formyl-6-methylquinoline

A201898-1G A201898-250MG A201898-25G
76800-384EA 16.98 USD
76800-384 76800-386 76800-388
2-Chloro-6-methylquinoline-3-carbaldehyde 97%
2-Chloro-6-methylquinoline-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₈ClNO
MW: 205.6400 g/mol
MDL Number: MFCD01912773
CAS Number: 73568-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR