Indocyanine green 98%

Supplier: Ambeed
Warning

Synonyms: IR-125, 4,5-benzoindotricarbocyanine, IC Green, Cardiogreen, Sodium 4-(2-(7-(1,1-dimethyl-3-(4-sulphonatobutyl)-1H-benzo[e]indol-2(3H)-ylidene)hepta-1,3,5-trien-1-yl)-1,1-dimethyl-1H-benzo[e]indol-3-ium-3-yl)butane-1-sulphonate, Foxgreen

A201360-100MG A201360-1G A201360-1MG A201360-250MG A201360-25MG A201360-5G
76800-366EA 42.44 USD
76800-366 76800-368 76800-370 76800-372 76800-374 76800-376
Indocyanine green 98%
Indocyanine green

Precaución: for Research Use Only

Formula: C₄₃H₄₇N₂NaO₆S₂
MW: 774.99 g/mol
MDL Number: MFCD00013078
CAS Number: 3599-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR