3-Methylbutyl isovalerate 98%

Supplier: Ambeed
Warning

Synonyms: Isovaleric acid Isopentyl ester, Isovaleric acid Isoamyl ester, Isoamyl Isovalerate, Isopentyl 3-methylbutanoate, Isopentyl Isovalerate

A201257-100G A201257-25G A201257-500G
76800-360EA 8.49 USD
76800-360 76800-362 76800-364
3-Methylbutyl isovalerate 98%
3-Methylbutyl isovalerate

Precaución: for Research Use Only

Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
MDL Number: MFCD00048369
CAS Number: 659-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR