4,5-Dichlorobenzene-1,3-disulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: Dichlorphenamide

A201176-10MG A201176-1MG A201176-50MG A201176-100MG A201176-1G A201176-250MG
76800-354EA 16.98 USD
76800-354 76800-356 76800-358 77626-000 77626-002 77626-004
4,5-Dichlorobenzene-1,3-disulfonamide 98%
4,5-Dichlorobenzene-1,3-disulfonamide

Precaución: for Research Use Only

Formula: C₆H₆Cl₂N₂O₄S₂
MW: 305.15 g/mol
MDL Number: MFCD00148948
CAS Number: 120-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR