Diethyl maleate 95%

Supplier: Ambeed
Warning

Synonyms: Maleic acid diethyl ester, Diethyl (Z)-2-butenedioate

A200510-1000G A200510-100G A200510-25G A200510-500G
76800-338EA 42 USD
76800-338 76800-340 76800-342 76800-344
Diethyl maleate 95%
Diethyl maleate

Precaución: for Research Use Only

Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Boiling Pt: 225
Melting Pt: -11
Flash Pt: 93
MDL Number: MFCD00009191
CAS Number: 141-05-9
Merck Index: 13,03150

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR