Ethyl-3-piperidinecarboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A200290-100G A200290-10G A200290-1G A200290-5G
76800-330EA 41 USD
76800-330 76800-332 76800-334 76800-336
Ethyl-3-piperidinecarboxylate hydrochloride 95%
Ethyl 3-piperidinecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₆ClNO₂
MDL Number: MFCD09032960
CAS Number: 4842-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR