Ethyl 4-amino-2-(methylthio)thiazole-5-carboxylate 95%

Supplier: Ambeed
Warning

A200198-1G A200198-250MG A200198-5G
76800-324EA 116.7 USD
76800-324 76800-326 76800-328
Ethyl 4-amino-2-(methylthio)thiazole-5-carboxylate 95%
Ethyl 4-amino-2-(methylthio)thiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂S₂
MW: 218.30 g/mol
MDL Number: MFCD01830310
CAS Number: 39736-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR