Fmoc-Ala(β-cyclobutyl)-OH 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-cyclobutylpropanoic acid, Fmoc-β-Cyclobutyl-Ala-OH

A200015-100MG A200015-1G A200015-250MG A200015-5G A200015-25G
76800-316EA 11 USD
76800-316 76800-318 76800-320 76800-322 77879-250
Fmoc-Ala(β-cyclobutyl)-OH 98%
Fmoc-Ala(β-cyclobutyl)-OH

Precaución: for Research Use Only

Formula: C₂₂H₂₃NO₄
MW: 365.422 g/mol
MDL Number: MFCD03840400
CAS Number: 478183-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR