(R)-Benzyl-2-amino-3-(4-hydroxyphenyl)propanoate-4-methylbenzenesulfonate 95%

Supplier: Ambeed
Warning

Synonyms: (R)-Benzyl 2-amino-3-(4-hydroxyphenyl)propanoate 4-methylbenzenesulphonate, H-D-Tyr-OBzl·p-tosylate

A199742-10G A199742-1G A199742-5G
76800-310EA 53.05 USD
76800-310 76800-312 76800-314
(R)-Benzyl-2-amino-3-(4-hydroxyphenyl)propanoate-4-methylbenzenesulfonate 95%
(R)-Benzyl-2-amino-3-(4-hydroxyphenyl)propanoate-4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₂₃H₂₅NO₆S
MW: 443.514 g/mol
MDL Number: MFCD00136658
CAS Number: 97984-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR