Fmoc-N-Me-Nle-OH 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Fmoc-N-methyl-L-norleucine
A199448-100G
A199448-10G
A199448-1G
A199448-25G
A199448-5G
76800-300EA
387
USD
76800-300
76800-302
76800-304
76800-306
76800-308
Fmoc-N-Me-Nle-OH 97%
Fmoc-N-Me-Nle-OH
Precaución: for Research Use Only
Formula:
C₂₂H₂₅NO₄ MW: 367.445 g/mol |
MDL Number:
MFCD00235878 CAS Number: 112883-42-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...