5,6-Dimethoxy-1H-indole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A198940-100MG A198940-1G A198940-250MG A198940-5G
76800-292EA 13.8 USD
76800-292 76800-294 76800-296 76800-298
5,6-Dimethoxy-1H-indole-2-carboxylic acid 95%
5,6-Dimethoxy-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₄
MW: 221.21 g/mol
MDL Number: MFCD00015459
CAS Number: 88210-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR