Ethyl-2-bromo-4-methylthiazole-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-bromo-4-methylthiazole-5-carboxylate

A198720-10G A198720-1G A198720-250MG A198720-25G A198720-5G
76800-282EA 28.65 USD
76800-282 76800-284 76800-286 76800-288 76800-290
Ethyl-2-bromo-4-methylthiazole-5-carboxylate 98%
Ethyl-2-bromo-4-methylthiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈BrNO₂S
MW: 250.12 g/mol
MDL Number: MFCD03791227
CAS Number: 22900-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR