4-(Ethoxymethyl)-2-methoxyphenol 97%

Supplier: Ambeed
Warning

A198590-100G A198590-10G A198590-1G A198590-25G A198590-5G
76800-272EA 183 USD
76800-272 76800-274 76800-276 76800-278 76800-280
4-(Ethoxymethyl)-2-methoxyphenol 97%
4-(Ethoxymethyl)-2-methoxyphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃
MDL Number: MFCD06409999
CAS Number: 13184-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR